Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082618.1 | NP_001076087.1 | 5, 6 | 593 | ||
NM_001772.3 | NP_001763.3 | 6, 7 | 974 | |||
XM_011527531.2 | XP_011525833.1 | 12, 13 | 2048 | |||
XM_011527532.2 | XP_011525834.1 | 11, 12 | 2011 | |||
XM_017027508.1 | XP_016882997.1 | 12, 13 | 2184 | |||
XM_017027509.1 | XP_016882998.1 | 10, 11 | 2088 | |||
XM_017027510.1 | XP_016882999.1 | 10, 11 | 1755 | |||
GenBank mRNA | AK090470.1 | - | 4 | 4000 | ||
AY162464.1 | - | 5, 6 | 561 | |||
BC028152.1 | - | 6, 7 | 939 | 5217182 | ||
CA426969.1 | - | 1 | 503 | |||
CA748646.1 | - | 1 | 500 |