Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098175.1 | NP_001091645.1 | 2 | 280 | ||
NM_001164178.1 | NP_001157650.1 | 2 | 180 | |||
NM_001164179.1 | NP_001157651.1 | 2 | 405 | |||
NM_001164182.1 | NP_001157654.1 | 2 | 405 | |||
NM_001164183.1 | NP_001157655.1 | 2 | 405 | |||
NM_001320916.1 | NP_001307845.1 | 2 | 180 | |||
NM_001776.5 | NP_001767.3 | 2 | 405 | |||
XM_011540371.2 | XP_011538673.1 | 3 | 416 | |||
XM_011540377.2 | XP_011538679.1 | 2 | 287 | |||
XM_017016958.1 | XP_016872447.1 | 2 | 547 | |||
XM_017016961.1 | XP_016872450.1 | 2 | 162 | |||
XM_017016963.1 | XP_016872452.1 | 2 | 172 | |||
XM_017016964.1 | XP_016872453.1 | 2 | 547 | |||
GenBank mRNA | AJ133133.1 | - | 2 | 285 | ||
AJ133134.1 | - | 2 | 285 | |||
AK301459.1 | - | 2 | 169 | |||
AK304018.1 | - | 2 | 166 | |||
AK304836.1 | - | 2 | 159 | |||
AL547722.3 | - | 2 | 162 | |||
BC047664.1 | - | 2 | 142 | 5762493 | ||
BF839210.1 | - | 2 | 189 | |||
CD652289.1 | - | 2 | 189 | |||
S73813.1 | - | 2 | 148 | |||
U87967.1 | - | 2 | 111 |