Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001248.3 | NP_001239.2 | 5 | 459 | ||
NM_001291960.1 | NP_001278889.1 | 5 | 500 | |||
NM_001291961.1 | NP_001278890.1 | 5 | 459 | |||
XM_011534266.2 | XP_011532568.1 | 5 | 459 | |||
GenBank mRNA | AF034840.2 | - | 5 | 420 | ||
AF039917.1 | - | 5 | 423 | |||
AK313322.1 | - | 5 | 485 | |||
BC029869.1 | - | 4 | 363 | 5174918 | ||
BX952692.1 | - | 5 | 434 | |||
DA209533.1 | - | 5 | 459 |