Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CD44 SIRNA Assays ›
Gene Symbol:
CD44
Gene Name:
CD44 molecule (Indian blood group)
Gene Aliases:
CDW44, CSPG8, ECMR-III, HCELL, HUTCH-I, IN, LHR, MC56, MDU2, MDU3, MIC4, Pgp1
Chromosome Location:
Chr.11: 35138870 - 35232402 on Build GRCh38
UniGene ID:
Hs.502328
Species:
Human
siRNA ID
HSS101596
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CD44
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000610.3
NP_000601.3
18
2555
NM_001001389.1
NP_001001389.1
17
2426
NM_001001390.1
NP_001001390.1
12
1808
NM_001001391.1
NP_001001391.1
9
1412
NM_001001392.1
NP_001001392.1
4
792
NM_001202555.1
NP_001189484.1
10
1616
NM_001202556.1
NP_001189485.1
8
1349
XM_005253231.2
XP_005253288.1
18
2285
XM_005253232.2
XP_005253289.1
18
2282
XM_005253233.2
XP_005253290.1
17
2165
XM_005253234.2
XP_005253291.1
17
2162
XM_005253235.2
XP_005253292.1
17
2156
XM_005253238.2
XP_005253295.1
14
1796
XM_005253239.2
XP_005253296.1
14
1790
XM_005253240.2
XP_005253297.1
13
1661
XM_006718388.1
XP_006718451.1
18
2279
XM_011520482.1
XP_011518784.1
17
2081
XM_011520483.1
XP_011518785.1
16
2030
XM_011520484.1
XP_011518786.1
16
2027
XM_011520485.1
XP_011518787.1
16
2024
XM_011520486.1
XP_011518788.1
13
1667
XM_011520488.1
XP_011518790.1
11
1331
XM_017018583.1
XP_016874072.1
17
2153
XM_017018584.1
XP_016874073.1
11
1394
XM_017018585.1
XP_016874074.1
10
1265
GenBank mRNA
AF098641.1
-
4
376
AJ251595.1
-
18
2299
AK129808.1
-
8
807
AK290424.1
-
9
978
AK297776.1
-
9
1018
AK304467.1
-
8
915
AK307707.1
-
8
873
AL832642.2
-
9
1412
AY101192.1
-
12
1490
AY101193.1
-
9
1095
BC004372.1
-
17
2123
3638681
BC052287.1
-
8
966
BC067348.1
-
9
1095
EF581837.1
-
10
1193
FJ216964.1
-
8
915
M24915.1
-
9
1093
M59040.1
-
9
1006
S66400.1
-
12
1424
U40373.1
-
9
1042
W47313.1
-
2
126
X55150.1
-
12
1497
X56794.1
-
12
1489
X66733.1
-
17
2121
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
cell-matrix adhesion
single organismal cell-cell adhesion
extracellular matrix disassembly
extracellular matrix organization
hyaluronan catabolic process
positive regulation of peptidyl-serine phosphorylation
positive regulation of heterotypic cell-cell adhesion
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of DNA damage response, signal transduction by p53 class mediator
cellular response to fibroblast growth factor stimulus
positive regulation of peptidyl-tyrosine phosphorylation
leukocyte migration
cartilage development
interferon-gamma-mediated signaling pathway
positive regulation of ERK1 and ERK2 cascade
monocyte aggregation
positive regulation of monocyte aggregation
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
cytokine receptor activity
protein binding
collagen binding
hyaluronic acid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline