Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000560.3 | NP_000551.1 | 4 | 473 | ||
NM_001040033.1 | NP_001035122.1 | 5 | 478 | |||
NM_001320638.1 | NP_001307567.1 | 4 | 473 | |||
GenBank mRNA | AK298723.1 | - | 4 | 420 | ||
AK313582.1 | - | 4 | 426 | |||
BC035456.1 | - | 5 | 460 | |||
BC040693.1 | - | 4 | 403 | 5498854 | ||
BG536920.1 | - | 4 | 474 | |||
BT019643.1 | - | 3 | 307 | |||
CA453414.1 | - | 480 | ||||
M37033.1 | - | 4 | 400 | |||
M60871.1 | - | 4 | 380 |