Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144822.1 | NP_001138294.1 | 4 | 789 | ||
NM_001779.2 | NP_001770.1 | 4 | 789 | |||
NR_026665.1 | 4 | 789 | ||||
XM_017002869.1 | XP_016858358.1 | 4 | 789 | |||
GenBank mRNA | AK291980.1 | - | 4 | 722 | ||
BC005930.1 | - | 4 | 735 | 4285827 | ||
BT019816.1 | - | 4 | 668 | |||
BT019817.1 | - | 4 | 668 | |||
CA312671.1 | - | 3 | 378 | |||
CD365881.1 | - | 3 | 413 | |||
CR456939.1 | - | 4 | 668 | |||
D28586.1 | - | 2 | 320 | |||
X06296.1 | - | 4 | 680 | |||
Y00636.1 | - | 4 | 677 |