Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000611.5 | NP_000602.1 | 3, 4 | 287 | ||
NM_001127223.1 | NP_001120695.1 | 2, 3 | 453 | |||
NM_001127225.1 | NP_001120697.1 | 3, 4 | 346 | |||
NM_001127226.1 | NP_001120698.1 | 3, 4 | 281 | |||
NM_001127227.1 | NP_001120699.1 | 3, 4 | 225 | |||
NM_203329.2 | NP_976074.1 | 4, 5 | 403 | |||
NM_203330.2 | NP_976075.1 | 5, 6 | 448 | |||
NM_203331.2 | NP_976076.1 | 4, 5 | 332 | |||
GenBank mRNA | AK128571.1 | - | 1, 2 | 2244 | ||
AK309894.1 | - | 2, 3 | 453 | |||
AK311778.1 | - | 3, 4 | 230 | |||
AL037045.3 | - | 3, 4 | 260 | |||
AL703204.1 | - | 4, 5 | 350 | |||
AU136899.1 | - | 4, 5 | 332 | |||
AU139009.1 | - | 3, 4 | 225 | |||
AU139541.1 | - | 3, 4 | 281 | |||
BC001506.2 | - | 3, 4 | 211 | 2988140 | ||
BI752924.1 | - | 5, 6 | 383 | |||
BQ230563.1 | - | 4, 5 | 251 | |||
BT007104.1 | - | 2, 3 | 171 | |||
CR407661.1 | - | 2, 3 | 171 | |||
CR541722.1 | - | 2, 3 | 171 | |||
DA250543.1 | - | 5, 6 | 394 | |||
DA578905.1 | - | 4, 5 | 347 | |||
DA840870.1 | - | 3, 4 | 347 | |||
DA917093.1 | - | 4, 5 | 314 | |||
M27909.1 | - | 3, 4 | 219 | |||
M34671.1 | - | 2, 3 | 200 | |||
M95708.1 | - | 3, 4 | 233 | |||
X16447.1 | - | 3, 4 | 234 | |||
X17198.1 | - | 3, 4 | 243 |