Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001782.2 | NP_001773.1 | 7, 8 | 1067 | ||
XM_017015351.1 | XP_016870840.1 | 8, 9 | 1440 | |||
GenBank mRNA | AB209587.1 | - | 5, 6 | 2859 | ||
AF283777.2 | - | 6, 7 | 1326 | |||
BC011734.2 | - | 7, 8 | 978 | |||
BC030227.1 | - | 7, 8 | 1008 | 5226648 | ||
DQ174771.1 | - | 8, 9 | 1436 | |||
M54992.1 | - | 7, 8 | 1051 |