Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001782.2 | NP_001773.1 | 8 | 1173 | ||
XM_017015351.1 | XP_016870840.1 | 9 | 1546 | |||
GenBank mRNA | AB209587.1 | - | 6 | 2965 | ||
AF283777.2 | - | 7 | 1432 | |||
BC011734.2 | - | 8 | 1084 | |||
BC030227.1 | - | 8 | 1114 | 5226648 | ||
DQ174771.1 | - | 9 | 1542 | |||
M54992.1 | - | 8 | 1157 |