Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001782.2 | NP_001773.1 | 5 | 516 | ||
XM_006716893.2 | XP_006716956.1 | 5 | 516 | |||
XM_006716894.3 | XP_006716957.1 | 6 | 889 | |||
XM_011518074.2 | XP_011516376.1 | 6 | 934 | |||
XM_017015351.1 | XP_016870840.1 | 6 | 889 | |||
GenBank mRNA | AB209587.1 | - | 3 | 2308 | ||
AF283777.2 | - | 4 | 775 | |||
AK307305.1 | - | 4 | 799 | |||
AK309268.1 | - | 3 | 1219 | |||
AK310093.1 | - | 5 | 483 | |||
AK310685.1 | - | 5 | 489 | |||
AK311283.1 | - | 5 | 765 | |||
BC011734.2 | - | 5 | 427 | |||
BC030227.1 | - | 5 | 457 | 5226648 | ||
DQ174771.1 | - | 6 | 885 | |||
M54992.1 | - | 5 | 500 |