Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199282.2 | NP_001186211.2 | 8 | 1255 | ||
NM_006726.4 | NP_006717.2 | 8 | 1214 | |||
XM_005263372.3 | XP_005263429.1 | 8 | 1400 | |||
XM_005263373.2 | XP_005263430.1 | 8 | 1442 | |||
XM_005263374.3 | XP_005263431.1 | 8 | 1399 | |||
XM_005263375.3 | XP_005263432.1 | 8 | 1399 | |||
XM_011532434.2 | XP_011530736.1 | 8 | 1399 | |||
XM_017008872.1 | XP_016864361.1 | 8 | 1442 | |||
GenBank mRNA | AF216648.2 | - | 8 | 1214 | ||
AF467287.1 | - | 8 | 1255 | |||
AK098552.1 | - | 7 | 828 |