Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001255.2 | NP_001246.2 | 9 | 1238 | ||
GenBank mRNA | AF099644.1 | - | 8 | 1181 | ||
AK225504.1 | - | 9 | 1217 | |||
AK303844.1 | - | 10 | 1193 | |||
AK312780.1 | - | 9 | 1217 | |||
BC000624.2 | - | 8 | 1245 | 3346682 | ||
BC001088.2 | - | 9 | 1238 | 2964686 | ||
BC006272.2 | - | 8 | 1180 | 3948819 | ||
BC009425.1 | - | 9 | 1220 | 2959460 | ||
BC009426.1 | - | 9 | 1220 | 2959596 | ||
BC010044.2 | - | 9 | 1229 | 3634656 | ||
BC012803.2 | - | 9 | 1220 | 2959588 | ||
BC012827.2 | - | 9 | 1238 | 2964686 | ||
BC013303.2 | - | 9 | 1220 | 2959588 | ||
BC015998.1 | - | 9 | 1216 | 4306426 | ||
BC024257.1 | - | 9 | 1220 | 2959460 | ||
BC031294.1 | - | 9 | 1220 | 2959487 | ||
BC110321.1 | - | 8 | 1201 | 5496957 | ||
BT007388.1 | - | 8 | 1137 | |||
U05340.1 | - | 9 | 1247 |