Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC25B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287516.1 | NP_001274445.1 | 13 | 1505 | ||
NM_001287517.1 | NP_001274446.1 | 13 | 1463 | |||
NM_001287518.1 | NP_001274447.1 | 12 | 1424 | |||
NM_001287519.1 | NP_001274448.1 | 13 | 1236 | |||
NM_001287520.1 | NP_001274449.1 | 13 | 1278 | |||
NM_001287522.1 | NP_001274451.1 | 12 | 1155 | |||
NM_001287524.1 | NP_001274453.1 | 9 | 1060 | |||
NM_004358.4 | NP_004349.1 | 13 | 2088 | |||
NM_021872.3 | NP_068658.1 | 12 | 2007 | |||
NM_021873.3 | NP_068659.1 | 13 | 2130 | |||
NR_136335.1 | 14 | 1357 | ||||
NR_136336.1 | 14 | 1584 | ||||
GenBank mRNA | AK092713.1 | - | 13 | 1278 | ||
AK295573.1 | - | 13 | 1347 | |||
AK299028.1 | - | 12 | 1231 | |||
AK299192.1 | - | 13 | 1281 | |||
BC006395.2 | - | 13 | 1411 | 4135465 | ||
BC009953.2 | - | 13 | 1491 | 4130804 | ||
BC051711.1 | - | 13 | 2035 | 5301205 | ||
BX640836.1 | - | 9 | 1052 | |||
BX647988.1 | - | 13 | 1193 | |||
FR695900.1 | - | 13 | 1278 | |||
FR695901.1 | - | 12 | 1155 | |||
M81934.1 | - | 13 | 1367 | |||
S78187.1 | - | 13 | 1535 | |||
Z68092.1 | - | 12 | 1315 |