Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308176.1 | NP_001295105.1 | 15 | 2499 | ||
NM_001792.4 | NP_001783.2 | 16 | 2979 | |||
XM_011525788.1 | XP_011524090.1 | 16 | 2644 | |||
GenBank mRNA | AK302831.1 | - | 15 | 2489 | ||
BC036470.1 | - | 16 | 2671 | 5264944 | ||
BG197310.1 | - | 1 | 44 | |||
M34064.1 | - | 13 | 2078 | |||
S42303.1 | - | 16 | 3013 | |||
X54315.1 | - | 16 | 2656 | |||
X57548.1 | - | 13 | 2081 |