Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290230.1 | NP_001277159.1 | 2, 3 | 477 | ||
NM_001798.4 | NP_001789.2 | 3, 4 | 555 | |||
NM_052827.3 | NP_439892.2 | 3, 4 | 555 | |||
XM_011537732.1 | XP_011536034.1 | 3, 4 | 555 | |||
GenBank mRNA | AB012305.1 | - | 3, 4 | 317 | ||
AK291941.1 | - | 3, 4 | 494 | |||
AK293246.1 | - | 2, 3 | 416 | |||
BC003065.2 | - | 3, 4 | 475 | 3504348 | ||
BJ991087.1 | - | 3, 4 | 552 | |||
BT006821.1 | - | 3, 4 | 315 | |||
M68520.1 | - | 3, 4 | 505 | |||
X61622.1 | - | 3, 4 | 315 | |||
X62071.1 | - | 3, 4 | 485 |