Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261.3 | NP_001252.1 | 4 | 486 | ||
XM_017014184.1 | XP_016869673.1 | 4 | 546 | |||
GenBank mRNA | AB451289.1 | - | 4 | 363 | ||
AB451421.1 | - | 4 | 363 | |||
AK313828.1 | - | 4 | 415 | |||
BC001968.1 | - | 4 | 423 | 3460767 | ||
BT019903.1 | - | 4 | 363 | |||
DB120461.1 | - | 4 | 486 | |||
L25676.1 | - | 4 | 427 | |||
X80230.1 | - | 4 | 461 |