Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001263.3 | NP_001254.2 | 10, 11 | 1539 | ||
XM_017007648.1 | XP_016863137.1 | 9, 10 | 1466 | |||
GenBank mRNA | AK314245.1 | - | 10, 11 | 1278 | ||
BC074833.2 | - | 10, 11 | 1090 | 30915545 | ||
BC074881.2 | - | 10, 11 | 1090 | 30915239 | ||
U60808.1 | - | 10, 11 | 1190 | |||
U65887.1 | - | 10, 11 | 1477 |