Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291484.2 | NP_001278413.1 | 5 | 1162 | ||
NM_001308398.1 | NP_001295327.1 | 5 | 1159 | |||
NM_004363.5 | NP_004354.3 | 5 | 1162 | |||
XM_005258413.3 | XP_005258470.2 | 5 | 1131 | |||
XM_017026145.1 | XP_016881634.1 | 5 | 1128 | |||
XM_017026146.1 | XP_016881635.1 | 4 | 876 | |||
GenBank mRNA | AB852566.1 | - | 3 | 481 | ||
AK223101.1 | - | 5 | 1098 | |||
BC034671.1 | - | 5 | 1110 | 5184800 | ||
M15042.1 | - | 5 | 1004 | |||
M16234.1 | - | 1 | 26 | |||
M17191.1 | - | 1 | 72 | |||
M29540.1 | - | 5 | 1129 | |||
X16455.1 | - | 5 | 1001 |