Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243645.1 | NP_001230574.1 | 3, 4 | 516 | ||
NM_021914.7 | NP_068733.1 | 3, 4 | 539 | |||
NM_138638.4 | NP_619579.1 | 3, 4 | 681 | |||
NR_028130.1 | 3, 4 | 538 | ||||
NR_028131.1 | 3, 4 | 427 | ||||
XM_011536363.2 | XP_011534665.1 | 3, 4 | 477 | |||
GenBank mRNA | AF087867.1 | - | 3, 4 | 434 | ||
AF134802.1 | - | 3, 4 | 550 | |||
AF134803.1 | - | 3, 4 | 444 | |||
AK314157.1 | - | 3, 4 | 499 | |||
AL599958.1 | - | 3, 4 | 515 | |||
BC011444.1 | - | 3, 4 | 465 | 4245104 | ||
BC022364.1 | - | 3, 4 | 548 | 4730117 | ||
BC022876.1 | - | 3, 4 | 552 | 4109108 | ||
BC025683.1 | - | 3, 4 | 427 | |||
CB992605.1 | - | 3, 4 | 381 | |||
DA298739.1 | - | 3, 4 | 504 | |||
DA897112.1 | - | 3, 4 | 515 |