Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000735.3 | NP_000726.1 | 4 | 464 | ||
NM_001252383.1 | NP_001239312.1 | 5 | 557 | |||
GenBank mRNA | AI200485.1 | - | 1 | 301 | ||
AK311972.1 | - | 4 | 421 | |||
BC010957.1 | - | 4 | 427 | 4043527 | ||
BC020782.1 | - | 4 | 423 | 4732686 | ||
BC055080.1 | - | 4 | 425 | 4044086 | ||
BG619154.1 | - | 4 | 466 | |||
CR457427.1 | - | 3 | 322 | |||
FJ624078.1 | - | 3 | 250 | |||
FJ624079.1 | - | 3 | 322 | |||
V00518.1 | - | 4 | 372 |