Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085471.1 | NP_001078940.1 | 2 | 493 | ||
NM_005197.3 | NP_005188.2 | 2 | 610 | |||
GenBank mRNA | AK308053.1 | - | 2 | 492 | ||
BC007506.2 | - | 2 | 504 | 3028787 | ||
U68723.1 | - | 2 | 488 |