Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085471.1 | NP_001078940.1 | 2, 3 | 672 | ||
NM_005197.3 | NP_005188.2 | 2, 3 | 789 | |||
GenBank mRNA | AK225967.1 | - | 4, 5 | 523 | ||
AK308053.1 | - | 2, 3 | 671 | |||
BC007506.2 | - | 2, 3 | 683 | 3028787 | ||
BP213901.1 | - | 542 | ||||
U68723.1 | - | 2, 3 | 667 |