Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHI3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276.2 | NP_001267.2 | 3 | 336 | ||
GenBank mRNA | AK130142.1 | - | 3 | 246 | ||
AK130200.1 | - | 3 | 246 | |||
AK225266.1 | - | 3 | 245 | |||
AK225584.1 | - | 3 | 245 | |||
AK312911.1 | - | 3 | 319 | |||
BC008568.1 | - | 3 | 282 | 4212748 | ||
BC038354.1 | - | 3 | 223 | 4212419 | ||
BC039132.1 | - | 3 | 232 | 4183798 | ||
BT007223.1 | - | 3 | 165 | |||
DA167112.1 | - | 3 | 341 | |||
M80927.1 | - | 3 | 236 |