Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025201.3 | NP_001020372.2 | 2, 3 | 355 | ||
NM_001822.5 | NP_001813.1 | 1, 2 | 354 | |||
GenBank mRNA | AK289941.1 | - | 1, 2 | 284 | ||
AK300890.1 | - | 2, 3 | 349 | |||
BC011393.1 | - | 1, 2 | 326 | 3871858 | ||
BQ893027.1 | - | 2, 3 | 388 | |||
DA516095.1 | - | 1, 2 | 364 | |||
DB453430.1 | - | 2, 3 | 418 | |||
Z22641.1 | - | 1, 2 | 23 |