Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025201.3 | NP_001020372.2 | 7 | 782 | ||
NM_001822.5 | NP_001813.1 | 6 | 781 | |||
GenBank mRNA | AK289941.1 | - | 6 | 711 | ||
AK300890.1 | - | 7 | 776 | |||
BC011393.1 | - | 6 | 753 | 3871858 | ||
Z22641.1 | - | 6 | 450 |