Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039936.2 | NP_001035025.1 | 5 | 1053 | ||
NM_001293069.1 | NP_001279998.1 | 12 | 1324 | |||
NM_001293070.1 | NP_001279999.1 | 12 | 1538 | |||
NM_001293071.1 | NP_001280000.1 | 10 | 1443 | |||
NM_001293072.1 | NP_001280001.1 | 11 | 1256 | |||
NM_001293073.1 | NP_001280002.1 | 5 | 918 | |||
NM_001293076.1 | NP_001280005.1 | 4 | 879 | |||
NM_001293081.1 | NP_001280010.1 | 5 | 568 | |||
NM_004067.3 | NP_004058.1 | 11 | 1499 | |||
NR_120525.1 | 3 | 659 | ||||
XM_011515105.2 | XP_011513407.1 | 14 | 1379 | |||
XM_011515106.2 | XP_011513408.1 | 13 | 1340 | |||
XM_011515107.2 | XP_011513409.1 | 13 | 2122 | |||
XM_017011721.1 | XP_016867210.1 | 15 | 1397 | |||
XM_017011722.1 | XP_016867211.1 | 14 | 2140 | |||
GenBank mRNA | AK026415.1 | - | 5 | 1053 | ||
AK293959.1 | - | 12 | 1118 | |||
AK294032.1 | - | 11 | 1256 | |||
AK294216.1 | - | 5 | 568 | |||
AK294271.1 | - | 12 | 1325 | |||
AK301235.1 | - | 5 | 577 | |||
AK313021.1 | - | 11 | 1273 | |||
AK316030.1 | - | 5 | 577 | |||
BC112155.1 | - | 11 | 1244 | 8327623 | ||
EU732751.1 | - | 3 | 260 | |||
EU732752.1 | - | 5 | 519 | |||
EU732754.1 | - | 4 | 480 | |||
EU732760.1 | - | 10 | 1006 | |||
EU732762.1 | - | 8 | 630 | |||
GQ924106.1 | - | 12 | 1325 | |||
U28926.1 | - | 11 | 1296 |