Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000744.6 | NP_000735.1 | 5 | 798 | ||
NM_001256573.1 | NP_001243502.1 | 5 | 729 | |||
NR_046317.1 | 5 | 823 | ||||
XM_011528524.1 | XP_011526826.1 | 4 | 442 | |||
XM_017027624.1 | XP_016883113.1 | 6 | 830 | |||
XM_017027625.1 | XP_016883114.1 | 6 | 848 | |||
GenBank mRNA | AK314907.1 | - | 5 | 595 | ||
BC096290.1 | - | 5 | 706 | 40007296 | ||
BC096291.1 | - | 6 | 614 | 40007297 | ||
BC096292.1 | - | 5 | 637 | 40007298 | ||
BC096293.3 | - | 5 | 731 | 40007299 | ||
L35901.1 | - | 5 | 745 | |||
U62433.1 | - | 5 | 798 | |||
X87629.1 | - | 4 | 492 | |||
X89745.1 | - | 1 | 282 | |||
Y08421.1 | - | 5 | 567 |