Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000745.3 | NP_000736.2 | 5 | 1335 | ||
GenBank mRNA | AF385586.1 | - | 5 | 1281 | ||
BC033639.1 | - | 5 | 1297 | 5501542 | ||
BC050469.1 | - | 1 | 747 | |||
M83712.1 | - | 5 | 1283 | |||
U62434.1 | - | 5 | 1289 | |||
Y08419.1 | - | 5 | 1135 |