Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000749.3 | NP_000740.1 | 5 | 521 | ||
XM_011544390.1 | XP_011542692.1 | 3 | 344 | |||
GenBank mRNA | BC069681.1 | - | 5 | 521 | 7262491 | |
BC069703.1 | - | 5 | 521 | 7262503 | ||
BC069788.1 | - | 5 | 521 | 7262515 | ||
U62438.1 | - | 5 | 489 | |||
X67513.1 | - | 4 | 288 | |||
Y08417.1 | - | 5 | 521 |