Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005199.4 | NP_005190.4 | 6 | 587 | ||
GenBank mRNA | AK125362.1 | - | 6 | 587 |