Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIRBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280.2 | NP_001271.1 | 5 | 565 | ||
NM_001300815.1 | NP_001287744.1 | 5 | 567 | |||
NM_001300829.1 | NP_001287758.1 | 5 | 567 | |||
NR_023312.2 | 5 | 626 | ||||
NR_023313.2 | 4 | 458 | ||||
XM_006722637.1 | XP_006722700.1 | 5 | 628 | |||
XM_011527668.1 | XP_011525970.1 | 5 | 501 | |||
XM_017026237.1 | XP_016881726.1 | 5 | 628 | |||
GenBank mRNA | AB451225.1 | - | 4 | 416 | ||
AB451348.1 | - | 4 | 416 | |||
AF021336.1 | - | 5 | 498 | |||
AK094781.1 | - | 4 | 391 | |||
AK095781.1 | - | 3 | 686 | |||
AK128423.1 | - | 4 | 480 | |||
AK304185.1 | - | 4 | 644 | |||
AK304470.1 | - | 5 | 399 | |||
BC000403.2 | - | 5 | 477 | 2822968 | ||
BC000901.1 | - | 5 | 482 | 2900585 | ||
BM706055.1 | - | 5 | 479 | |||
BP195090.1 | - | 5 | 565 | |||
BT007210.1 | - | 4 | 416 | |||
D78134.1 | - | 5 | 496 | |||
DA433715.1 | - | 5 | 502 |