Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281.2 | NP_001272.2 | 6 | 1296 | ||
NM_001300971.1 | NP_001287900.1 | 6 | 708 | |||
GenBank mRNA | AF013488.1 | - | 6 | 786 | ||
AI126083.1 | - | 1 | 187 | |||
AI148860.1 | - | 1 | 189 | |||
BC005969.1 | - | 6 | 773 | 4076810 | ||
BC052812.1 | - | 6 | 796 | 6527446 | ||
BQ923739.1 | - | 6 | 711 | |||
CR541971.1 | - | 5 | 568 | |||
D49738.1 | - | 6 | 841 |