Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP2M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025205.1 | NP_001020376.1 | 7 | 960 | ||
NM_001311198.1 | NP_001298127.1 | 10 | 1041 | |||
NM_004068.3 | NP_004059.2 | 8 | 966 | |||
GenBank mRNA | AK057883.1 | - | 8 | 957 | ||
AK294159.1 | - | 8 | 787 | |||
AK295999.1 | - | 8 | 750 | |||
AK297578.1 | - | 7 | 641 | |||
AK297849.1 | - | 10 | 1032 | |||
AK300228.1 | - | 4 | 522 | |||
AK304510.1 | - | 8 | 892 | |||
BC004996.1 | - | 8 | 944 | 2958005 | ||
BC013796.1 | - | 7 | 878 | 3924339 | ||
BC014030.1 | - | 8 | 934 | 4541249 | ||
BT007308.1 | - | 7 | 809 | |||
D63475.1 | - | 8 | 895 | |||
U36188.1 | - | 7 | 855 |