Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289.5 | NP_001280.3 | 6 | 927 | ||
GenBank mRNA | AK292785.1 | - | 6 | 860 | ||
BC005367.1 | - | 6 | 803 | |||
BC022305.1 | - | 6 | 803 | 4663624 | ||
Y12696.1 | - | 6 | 894 |