Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162407.1 | NP_001155879.1 | 10, 11 | 1601 | ||
NM_004071.3 | NP_004062.2 | 10, 11 | 1321 | |||
NR_027855.1 | 9, 10 | 1230 | ||||
NR_027856.1 | 8, 9 | 2721 | ||||
GenBank mRNA | AK025306.1 | - | 5, 6 | 2628 | ||
AK294295.1 | - | 10, 11 | 1601 | |||
AK316127.1 | - | 10, 11 | 1292 | |||
BC028149.1 | - | 9, 10 | 1126 | |||
BC031549.1 | - | 10, 11 | 1217 | 4387232 | ||
L29219.1 | - | 10, 11 | 1295 | |||
L29222.1 | - | 9, 10 | 1204 | |||
M59287.1 | - | 9, 10 | 1205 |