Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005508.4 | NP_005499.1 | 2 | 467 | ||
XM_017005687.1 | XP_016861176.1 | 2 | 514 | |||
GenBank mRNA | AK292674.1 | - | 2 | 397 | ||
BC069139.1 | - | 1 | 352 | 7216998 | ||
BC071751.1 | - | 2 | 467 | 30415446 | ||
BC074935.2 | - | 1 | 327 | 30915268 | ||
CD639869.1 | - | 2 | 496 | |||
CD640816.1 | - | 2 | 470 |