Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256063.1 | NP_001242992.1 | 7 | 841 | ||
NM_001256064.1 | NP_001242993.1 | 7 | 1054 | |||
NM_001843.3 | NP_001834.2 | 7 | 841 | |||
NM_175038.2 | NP_778203.1 | 5 | 657 | |||
XM_005268651.2 | XP_005268708.1 | 7 | 1204 | |||
XM_006719241.2 | XP_006719304.1 | 7 | 828 | |||
XM_011537926.2 | XP_011536228.1 | 7 | 793 | |||
XM_011537927.2 | XP_011536229.1 | 7 | 850 | |||
XM_017018821.1 | XP_016874310.1 | 7 | 844 | |||
XM_017018822.1 | XP_016874311.1 | 7 | 793 | |||
XM_017018823.1 | XP_016874312.1 | 7 | 1204 | |||
XM_017018824.1 | XP_016874313.1 | 7 | 828 | |||
XM_017018825.1 | XP_016874314.1 | 7 | 793 | |||
XM_017018826.1 | XP_016874315.1 | 7 | 828 | |||
XM_017018827.1 | XP_016874316.1 | 7 | 850 | |||
GenBank mRNA | AK289544.1 | - | 7 | 933 | ||
AK289698.1 | - | 7 | 783 | |||
BC036569.2 | - | 7 | 780 | 5273941 | ||
U07819.1 | - | 6 | 614 | |||
U07820.1 | - | 5 | 662 | |||
Z21488.1 | - | 7 | 735 |