Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000088.3 | NP_000079.2 | 49 | 3981 | ||
XM_005257058.3 | XP_005257115.2 | 47 | 3720 | |||
XM_005257059.4 | XP_005257116.2 | 36 | 3072 | |||
XM_011524341.1 | XP_011522643.1 | 46 | 3792 | |||
GenBank mRNA | AB209597.1 | - | 49 | 3956 | ||
AK297731.1 | - | 6 | 1104 | |||
BC036531.2 | - | 49 | 3971 | 5264710 | ||
JQ236861.1 | - | 49 | 3847 | |||
K01228.1 | - | 31 | 2583 | |||
S64596.1 | - | 2 | 321 | |||
X06269.1 | - | 2 | 171 | |||
Z74615.1 | - | 49 | 3974 |