Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000088.3 | NP_000079.2 | 49, 50 | 4122 | ||
XM_005257058.3 | XP_005257115.2 | 47, 48 | 3861 | |||
XM_005257059.4 | XP_005257116.2 | 36, 37 | 3213 | |||
XM_011524341.1 | XP_011522643.1 | 46, 47 | 3933 | |||
GenBank mRNA | AB209597.1 | - | 49, 50 | 4097 | ||
BC036531.2 | - | 49, 50 | 4112 | 5264710 | ||
JQ236861.1 | - | 49, 50 | 3988 | |||
K01228.1 | - | 31, 32 | 2724 | |||
X06269.1 | - | 2, 3 | 312 | |||
Z74615.1 | - | 49, 50 | 4115 |