Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000089.3 | NP_000080.2 | 49 | 3792 | ||
GenBank mRNA | AK226074.1 | - | 49 | 3443 | ||
AL833478.1 | - | 6 | 4100 | |||
BC042586.1 | - | 49 | 3439 | 4803351 | ||
BC054498.1 | - | 49 | 3421 | 6474549 | ||
J03464.1 | - | 49 | 3790 | |||
KU177988.1 | - | 49 | 3321 | |||
V00503.1 | - | 18 | 1456 | |||
X55525.1 | - | 6 | 446 | |||
Z74616.1 | - | 49 | 3460 |