Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000090.3 | NP_000081.1 | 2 | 226 | ||
GenBank mRNA | AK308010.1 | - | 2 | 226 | ||
BC028178.1 | - | 2 | 224 | 5405119 | ||
BP374999.1 | - | 2 | 226 | |||
X07240.1 | - | 2 | 109 | |||
X14420.1 | - | 2 | 211 |