Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001845.5 | NP_001836.3 | 30 | 2342 | ||
XM_011521048.2 | XP_011519350.1 | 30 | 10274 | |||
GenBank mRNA | BC047305.1 | - | 5 | 431 | ||
BC151220.1 | - | 30 | 2326 | 40148649 | ||
X05561.1 | - | 30 | 2334 | |||
Y00706.1 | - | 28 | 2077 |