Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL5A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000393.3 | NP_000384.2 | 1, 2 | 378 | ||
GenBank mRNA | AB209045.1 | - | 1, 2 | 240 | ||
AK297936.1 | - | 1, 2 | 260 | |||
AK300146.1 | - | 1, 2 | 266 | |||
AL552427.3 | - | 1, 2 | 378 | |||
BC086874.1 | - | 250 | 30708812 | |||
J04478.1 | - | 1, 2 | 241 | |||
Y14690.1 | - | 1, 2 | 241 |