Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL7A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000094.3 | NP_000085.1 | 23 | 3021 | ||
XM_011533337.1 | XP_011531639.1 | 24 | 3056 | |||
XM_017005688.1 | XP_016861177.1 | 24 | 3056 | |||
XM_017005689.1 | XP_016861178.1 | 24 | 3056 | |||
XM_017005690.1 | XP_016861179.1 | 24 | 3056 | |||
XM_017005691.1 | XP_016861180.1 | 24 | 3056 | |||
XM_017005692.1 | XP_016861181.1 | 24 | 3056 | |||
GenBank mRNA | D13694.1 | - | 19 | 2435 | ||
L02870.1 | - | 23 | 3133 | |||
M65158.1 | - | 5 | 578 | |||
M96984.1 | - | 15 | 1916 | |||
S51236.1 | - | 15 | 1916 |