Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL9A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001853.3 | NP_001844.3 | 4, 5 | 262 | ||
XM_011528545.1 | XP_011526847.1 | 4, 5 | 260 | |||
XM_017027666.1 | XP_016883155.1 | 4, 5 | 260 | |||
GenBank mRNA | BC011705.2 | - | 4, 5 | 262 | 3830925 | |
L41162.1 | - | 4, 5 | 259 |