Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL17A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000494.3 | NP_000485.3 | 30, 31 | 2440 | ||
GenBank mRNA | AB900157.1 | - | 29, 30 | 2271 | ||
M63730.1 | - | 13, 14 | 750 | |||
M91669.1 | - | 30, 31 | 2376 |