Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL17A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000494.3 | NP_000485.3 | 6, 7 | 542 | ||
GenBank mRNA | AB900157.1 | - | 5, 6 | 373 | ||
AK309940.1 | - | 6, 7 | 488 | |||
BC004478.2 | - | 6, 7 | 524 | |||
M91669.1 | - | 6, 7 | 478 |