Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162861.2 | NP_001156333.1 | 2 | 575 | ||
NM_001162862.2 | NP_001156334.1 | 2 | 575 | |||
NM_001321518.1 | NP_001308447.1 | 2 | 575 | |||
NM_004375.4 | NP_004366.1 | 2 | 575 | |||
NR_027941.2 | 2 | 575 | ||||
NR_027942.2 | 2 | 575 | ||||
NR_135677.1 | 2 | 575 | ||||
XM_011524342.2 | XP_011522644.1 | 2 | 2814 | |||
XM_017024192.1 | XP_016879681.1 | 2 | 2814 | |||
XM_017024193.1 | XP_016879682.1 | 2 | 2814 | |||
XM_017024194.1 | XP_016879683.1 | 2 | 570 | |||
XM_017024195.1 | XP_016879684.1 | 2 | 570 | |||
XM_017024196.1 | XP_016879685.1 | 2 | 570 | |||
GenBank mRNA | AF044321.1 | - | 2 | 451 | ||
AI828544.1 | - | 4 | 610 | |||
AK293851.1 | - | 2 | 408 | |||
AK295381.1 | - | 2 | 493 | |||
BC005895.1 | - | 2 | 446 | 4282744 | ||
BF725369.1 | - | 2 | 575 | |||
BI598312.1 | - | 2 | 491 | |||
BM975167.1 | - | 4 | 643 | |||
CR541837.1 | - | 2 | 436 | |||
U79270.1 | - | 1 | 66 |