Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001868.3 | NP_001859.1 | 8 | 1093 | ||
GenBank mRNA | AK124421.1 | - | 1 | 1521 | ||
AK131054.1 | - | 471 | ||||
AK291493.1 | - | 8 | 970 | |||
BC005279.1 | - | 8 | 965 | 3949850 | ||
BT007313.1 | - | 8 | 943 | |||
X67318.1 | - | 8 | 950 |