Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001870.3 | NP_001861.2 | 9, 10 | 1037 | ||
GenBank mRNA | AK313628.1 | - | 9, 10 | 997 | ||
BC012613.1 | - | 9, 10 | 1000 | 4104766 | ||
BF508919.1 | - | 2, 3 | 659 | |||
M27717.1 | - | 9, 10 | 985 | |||
S40234.1 | - | 6, 7 | 658 |